Merck
  • Mild intrauterine hypoperfusion reproduces neurodevelopmental disorders observed in prematurity.
Mild intrauterine hypoperfusion reproduces neurodevelopmental disorders observed in prematurity.
Scientific reports (2016-12-21)
Makiko Ohshima, Jacques-Olivier Coq, Kentaro Otani, Yorito Hattori, Yuko Ogawa, Yoshiaki Sato, Mariko Harada-Shiba, Masafumi Ihara, Masahiro Tsuji
ABSTRACT
Severe intrauterine ischemia is detrimental to the developing brain. The impact of mild intrauterine hypoperfusion on neurological development, however, is still unclear. We induced mild intrauterine hypoperfusion in rats on embryonic day 17 via arterial stenosis with metal microcoils wrapped around the uterine and ovarian arteries. All pups were born with significantly decreased birth weights. Decreased gray and white matter areas were observed without obvious tissue damage. Pups presented delayed newborn reflexes, muscle weakness, and altered spontaneous activity. The levels of proteins indicative of inflammation and stress in the vasculature, i.e., RANTES, vWF, VEGF, and adiponectin, were upregulated in the placenta. The levels of mRNA for proteins associated with axon and astrocyte development were downregulated in fetal brains. The present study demonstrates that even mild intrauterine hypoperfusion can alter neurological development, which mimics the clinical signs and symptoms of children with neurodevelopmental disorders born prematurely or with intrauterine growth restriction.
Product Description
Millipore
MILLIPLEX® Rat Vascular Injury Magnetic Bead Panel 2 - Toxicity Multiplex Assay, The analytes available for this multiplex kit are: Adiponectin, sE-Selectin, and sICAM-1.
Millipore
MILLIPLEX® Rat Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in rat serum, plasma and cell culture samples.