Merck
  • Dipeptide-functionalized polyamidoamine dendrimer-mediated apoptin gene delivery facilitates apoptosis of human primary glioma cells.
Dipeptide-functionalized polyamidoamine dendrimer-mediated apoptin gene delivery facilitates apoptosis of human primary glioma cells.
International journal of pharmaceutics (2016-11-05)
Yoonhee Bae, Eric S Green, Goo-Young Kim, Su Jeong Song, Ji Young Mun, Sunray Lee, Jong-Il Park, Jong-Sang Park, Kyung Soo Ko, Jin Han, Joon Sig Choi
ABSTRACT
Glioblastoma multiform (GBM) is the most frequent and aggressive form of brain tumors in adults. However, the development of more efficient and safe nonviral vector gene therapy represents a promising therapeutic approach, using a tumor-specific killer gene, named apoptin. In this study, we describe the efficacy of non-viral gene delivery vectors, the amino acid-conjugated PAMAM derivatives (PAMAM-H-R and PAMAM-H-K) in delivering a therapeutic gene, displaying affinity toward human primary glioma cells (GBL-14 cells) and dermal fibroblasts. We analyzed transfection efficiency, using luciferase (Luci) and a pDNA encoding for enhanced fluorescent protein (EGFP), and cytotoxicity in both cells. The results show that transfection efficiency of PAMAM-H-R improved compared to native PAMAM dendrimer, but cytotoxicity of PAMAM-H-R and PAMAM-H-K were very low. We treated both cells with a polyplex formation of PAMAM-H-R or PAMAM-H-K/apoptin, and analyzed their cellular uptake and localization by flow cytometry and confocal microscopy. Furthermore, we analyzed the endosomal escape effect using TEM images, and found that PAMAM-H-R showed very fast escape from endosome to the cytosol. Caspase 3 activity assay, cell cycle distribution, and JC-1 analysis showed apoptosis induced by apoptin in GBL-14 cells. This indicates that PAMAM-H-R can be a potential nonviral vector gene delivery carrier for brain tumor therapy. The present study demonstrates that PAMAM-H-R/apoptin gene polyplex can be used as an effective therapeutic candidate for GBM due to its selective induction of apoptosis in primary glioma cells as a potential nonviral gene delivery carrier for brain tumor therapy.
Sigma-Aldrich
Fmoc-His(Trt)-OH, ≥98.0% (sum of enantiomers, HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
HBTU, ≥98.0% (T)
Pricing and availability is not currently available.