Merck
  • A covalent and cleavable antibody-DNA conjugation strategy for sensitive protein detection via immuno-PCR.
A covalent and cleavable antibody-DNA conjugation strategy for sensitive protein detection via immuno-PCR.
Scientific reports (2016-03-08)
Jessie A G L van Buggenum, Jan P Gerlach, Selma Eising, Lise Schoonen, Roderick A P M van Eijl, Sabine E J Tanis, Mark Hogeweg, Nina C Hubner, Jan C van Hest, Kimberly M Bonger, Klaas W Mulder
ABSTRACT
Immuno-PCR combines specific antibody-based protein detection with the sensitivity of PCR-based quantification through the use of antibody-DNA conjugates. The production of such conjugates depends on the availability of quick and efficient conjugation strategies for the two biomolecules. Here, we present an approach to produce cleavable antibody-DNA conjugates, employing the fast kinetics of the inverse electron-demand Diels-Alder reaction between tetrazine and trans-cyclooctene (TCO). Our strategy consists of three steps. First, antibodies are functionalized with chemically cleavable NHS-s-s-tetrazine. Subsequently, double-stranded DNA is functionalized with TCO by enzymatic addition of N3-dATP and coupling to trans-Cyclooctene-PEG12-Dibenzocyclooctyne (TCO-PEG12-DBCO). Finally, conjugates are quickly and efficiently obtained by mixing the functionalized antibodies and dsDNA at low molar ratios of 1:2. In addition, introduction of a chemically cleavable disulphide linker facilitates release and sensitive detection of the dsDNA after immuno-staining. We show specific and sensitive protein detection in immuno-PCR for human epidermal stem cell markers, ITGA6 and ITGB1, and the differentiation marker Transglutaminase 1 (TGM1). We anticipate that the production of chemically cleavable antibody-DNA conjugates will provide a solid basis for the development of multiplexed immuno-PCR experiments and immuno-sequencing methodologies.
Sigma-Aldrich
Monoclonal Anti-AGO2 antibody produced in rat, ~1.5 mg/mL, clone 11A9, purified immunoglobulin
Pricing and availability is not currently available.