Merck
  • Cancer cell death induced by novel small molecules degrading the TACC3 protein via the ubiquitin-proteasome pathway.
Cancer cell death induced by novel small molecules degrading the TACC3 protein via the ubiquitin-proteasome pathway.
Cell death & disease (2014-11-07)
N Ohoka, K Nagai, T Hattori, K Okuhira, N Shibata, N Cho, M Naito
ABSTRACT
The selective degradation of target proteins with small molecules is a novel approach to the treatment of various diseases, including cancer. We have developed a protein knockdown system with a series of hybrid small compounds that induce the selective degradation of target proteins via the ubiquitin-proteasome pathway. In this study, we designed and synthesized novel small molecules called SNIPER(TACC3)s, which target the spindle regulatory protein transforming acidic coiled-coil-3 (TACC3). SNIPER(TACC3)s induce poly-ubiquitylation and proteasomal degradation of TACC3 and reduce the TACC3 protein level in cells. Mechanistic analysis indicated that the ubiquitin ligase APC/C(CDH1) mediates the SNIPER(TACC3)-induced degradation of TACC3. Intriguingly, SNIPER(TACC3) selectively induced cell death in cancer cells expressing a larger amount of TACC3 protein than normal cells. These results suggest that protein knockdown of TACC3 by SNIPER(TACC3) is a potential strategy for treating cancers overexpressing the TACC3 protein.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys48-Specific, clone Apu2, rabbit monoclonal, clone Apu2, from rabbit
Pricing and availability is not currently available.