- Alkaline Cytosolic pH and High Sodium Hydrogen Exchanger 1 (NHE1) Activity in Th9 Cells.
Alkaline Cytosolic pH and High Sodium Hydrogen Exchanger 1 (NHE1) Activity in Th9 Cells.
The Journal of biological chemistry (2016-09-16)
Yogesh Singh, Yuetao Zhou, Xiaolong Shi, Shaqiu Zhang, Anja T Umbach, Madhuri S Salker, Karl S Lang, Florian Lang
PMID27629415
ABSTRACT
CD4