- Performance of Broiler Chickens Fed Low Protein, Limiting Amino Acid Supplemented Diets Formulated Either on Total or Standardized Ileal Digestible Amino Acid Basis.
Performance of Broiler Chickens Fed Low Protein, Limiting Amino Acid Supplemented Diets Formulated Either on Total or Standardized Ileal Digestible Amino Acid Basis.
The aim of present experiment was to investigate the effect of protein reduction in commercial broiler chicken rations with incorporation of de-oiled rice bran (DORB) and supplementation of limiting amino acids (valine, isoleucine, and/or tryptophan) with ration formulation either on total amino acid (TAA) or standardized ileal digestible amino acids (SIDAA). The experimental design consisted of T