Merck
  • Liprin beta 1, a member of the family of LAR transmembrane tyrosine phosphatase-interacting proteins, is a new target for the metastasis-associated protein S100A4 (Mts1).
Liprin beta 1, a member of the family of LAR transmembrane tyrosine phosphatase-interacting proteins, is a new target for the metastasis-associated protein S100A4 (Mts1).
The Journal of biological chemistry (2002-02-12)
Marina Kriajevska, Margrethe Fischer-Larsen, Ejvind Moertz, Ole Vorm, Eugene Tulchinsky, Mariam Grigorian, Noona Ambartsumian, Eugene Lukanidin
ABSTRACT
Metastasis-associated protein S100A4 (Mts1) induces invasiveness of primary tumors and promotes metastasis. S100A4 belongs to the family of small calcium-binding S100 proteins that are involved in different cellular processes as transducers of calcium signal. S100A4 modulates properties of tumor cells via interaction with its intracellular targets, heavy chain of non-muscle myosin and p53. Here we report identification of a new molecular target of the S100A4 protein, liprin beta1. Liprin beta1 belongs to the family of leukocyte common antigen-related (LAR) transmembrane tyrosine phosphatase-interacting proteins that may regulate LAR protein properties via interaction with another member of the family, liprin alpha1. We showed by the immunoprecipitation analysis that S100A4 interacts specifically with liprin beta1 in vivo. Immunofluorescence staining demonstrated the co-localization of S100A4 and liprin beta1 in the cytoplasm and particularly at the protrusion sites of the plasma membrane. We mapped the S100A4 binding site at the C terminus of the liprin beta1 molecule between amino acid residues 938 and 1005. The S100A4-binding region contains two putative phosphorylation sites by protein kinase C and protein kinase CK2. S100A4-liprin beta1 interaction resulted in the inhibition of liprin beta1 phosphorylation by both kinases in vitro.
Sigma-Aldrich
Anti-Chicken IgY (IgG) (whole molecule) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.