Merck
Kaempferitrin prevents bone lost in ovariectomized rats.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2015-11-26)
Xue-Qin Ma, Ting Han, Xia Zhang, Jin-Zhong Wu, Khalid Rahman, Lu-Ping Qin, Cheng-Jian Zheng
ABSTRACT
Podocarpium podocarpum (DC.), an edible and medicinal plant popularly used for the treatment of bruises and fracture in Chinese folk medicine, has been proved to possess significant antiosteoporotic effect in our latest research. Our study aimed to investigate the in vitro and in vivo antiosteoporotic effect of kaempfertrin (KN), a principal flavonoid in P. podocarpum obtained through bio-guided isolation. An ovariectomized (OVX) rat model of osteoporosis as well as in vitro osteoblast and osteoclast cell lines were employed to evaluate the antiosteoporotic potency of KN. KN significantly improved the bone mass and microarchitecture in OVX rats, with little estrogen-like side effect compared with estradiol valerate. KN also exhibited stimulatory effect on osteoblastic cells and inhibitory action on osteoclastic cells, which down-regulated the phosphorylation level of I-κB. KN possessed significant antiosteoporotic activity. Combined with its limited estrogen-like side effect, KN can be regarded as an idealistic antiosteoporotic candidate for human osteoporosis diseases.
Sigma-Aldrich
Kaempferitrin, ≥97% (NMR)
Pricing and availability is not currently available.