Merck
  • Zebularine-induced reduction in VEGF secretion by HIF-1α degradation in oral squamous cell carcinoma.
Zebularine-induced reduction in VEGF secretion by HIF-1α degradation in oral squamous cell carcinoma.
Molecular medicine reports (2008-07-01)
Maiko Suzuki, Fumiaki Shinohara, Hidemi Rikiishi
ABSTRACT
Vascular endothelial growth factor (VEGF) is a potent inducer of angiogenesis in oral squamous cell carcinoma (OSCC). In this study, we used the novel DNA methyltransferase inhibitor zebularine (Zeb) to investigate epigenetic influences on the secretion of VEGF-A in the OSCC cell line HSC-3. Under normoxic conditions, we found that Zeb inhibited secretion in a dose-dependent manner by reducing the activity of hypoxia-inducible factor-1α (HIF-1α). Treatment of the cells with the proteasome inhibitor MG132 protected the HIF-1α protein from Zeb-mediated epigenetic regulation. In addition, our study revealed that neither the PI3K/Akt nor the p53 signaling pathway is required for Zeb-induced HIF-1α degradation. In short, Zeb influenced the stability of the HIF-1α protein and the activity of its targets, such as VEGF, in HSC-3 cells in normoxic conditions. This study has laid the foundation for a novel anti-cancer approach, which may find applications in molecular staging.
Sigma-Aldrich
Imprint® Methylated DNA Quantification Kit, To measure global DNA methylation shifts from as low as 10 ng DNA