Merck
  • A single-headed fission yeast myosin V transports actin in a tropomyosin-dependent manner.
A single-headed fission yeast myosin V transports actin in a tropomyosin-dependent manner.
The Journal of cell biology (2016-07-20)
Qing Tang, Neil Billington, Elena B Krementsova, Carol S Bookwalter, Matthew Lord, Kathleen M Trybus
ABSTRACT
Myo51, a class V myosin in fission yeast, localizes to and assists in the assembly of the contractile ring, a conserved eukaryotic actomyosin structure that facilitates cytokinesis. Rng8 and Rng9 are binding partners that dictate the cellular localization and function of Myo51. Myo51 was expressed in insect cells in the presence or absence of Rng8/9. Surprisingly, electron microscopy of negatively stained images and hydrodynamic measurements showed that Myo51 is single headed, unlike most class V myosins. When Myo51-Rng8/9 was bound to actin-tropomyosin, two attachment sites were observed: the typical ATP-dependent motor domain attachment and a novel ATP-independent binding of the tail mediated by Rng8/9. A modified motility assay showed that this additional binding site anchors Myo51-Rng8/9 so that it can cross-link and slide actin-tropomyosin filaments relative to one another, functions that may explain the role of this motor in contractile ring assembly.
Sigma-Aldrich
Met-Ala-Ser
Pricing and availability is not currently available.