Merck
  • New class of nucleophiles for palladium-catalyzed asymmetric allylic alkylation. Total synthesis of agelastatin A.
New class of nucleophiles for palladium-catalyzed asymmetric allylic alkylation. Total synthesis of agelastatin A.
Journal of the American Chemical Society (2006-05-04)
Barry M Trost, Guangbin Dong
ABSTRACT
New classes of nucleophiles, pyrroles, and N-methoxyamides were developed for Pd-catalyzed AAA reactions. By varying the functional groups at the 2-position of pyrroles, either regioisomer of the piperazinone is available. Using one regioisomer, the total synthesis of (+)-agelastatin A in 10 total steps is accomplished. For this synthesis, a new copper-catalyzed aziridination and an indium-catalyzed oxidative ring opening of a N-tosylaziridine were developed. The feasibility of accessing (-)-agelastatin A from the same enantiomer of the chiral catalyst from the other regioisomeric piperazinone is indicated.
Sigma-Aldrich
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I)
Pricing and availability is not currently available.