- Novel synthesis of 2-chloroquinolines from 2-vinylanilines in nitrile solvent.
Novel synthesis of 2-chloroquinolines from 2-vinylanilines in nitrile solvent.
The Journal of organic chemistry (2002-10-26)
Byoung Se Lee, Jae Hak Lee, Dae Yoon Chi
PMID12398521
ABSTRACT
2-Vinyl- or heteroaryl-substituted anilines were reacted with diphosgene in acetonitrile solution via a reactive imidoyl moiety to afford the corresponding 2-chloroquinolines. Facile syntheses of nine 2-chloroquinoline derivatives from several anilines and their postulate mechanism is described. The postulate mechanism of 2-chloroquinoline formation via imidoyl moiety as a good leaving group shows that the reaction consists of the following three steps: (1) generation of phenylisocyanate, (2) quinoline ring formation, and (3) chlorination on C2 position of quinoline.
MATERIALS