Merck
Isolation and culture of adult epithelial stem cells from human skin.
Journal of visualized experiments : JoVE (2011-04-15)
Zhiru Guo, Kyle Draheim, Stephen Lyle
ABSTRACT
The homeostasis of all self-renewing tissues is dependent on adult stem cells. As undifferentiated stem cells undergo asymmetric divisions, they generate daughter cells that retain the stem cell phenotype and transit-amplifying cells (TA cells) that migrate from the stem cell niche, undergo rapid proliferation and terminally differentiate to repopulate the tissue. Epithelial stem cells have been identified in the epidermis, hair follicle, and intestine as cells with a high in vitro proliferative potential and as slow-cycling label-retaining cells in vivo (1-3). Adult, tissue-specific stem cells are responsible for the regeneration of the tissues in which they reside during normal physiologic turnover as well as during times of stress (4-5). Moreover, stem cells are generally considered to be multi-potent, possessing the capacity to give rise to multiple cell types within the tissue (6). For example, rodent hair follicle stem cells can generate epidermis, sebaceous glands, and hair follicles (7-9). We have shown that stem cells from the human hair follicle bulge region exhibit multi-potentiality (10). Stem cells have become a valuable tool in biomedical research, due to their utility as an in vitro system for studying developmental biology, differentiation, tumorigenesis and for their possible therapeutic utility. It is likely that adult epithelial stem cells will be useful in the treatment of diseases such as ectodermal dysplasias, monilethrix, Netherton syndrome, Menkes disease, hereditary epidermolysis bullosa and alopecias (11-13). Additionally, other skin problems such as burn wounds, chronic wounds and ulcers will benefit from stem cell related therapies (14,15). Given the potential for reprogramming of adult cells into a pluripotent state (iPS cells)(16,17), the readily accessible and expandable adult stem cells in human skin may provide a valuable source of cells for induction and downstream therapy for a wide range of disease including diabetes and Parkinson's disease.
Product Description
Roche
Transferrin, from human serum
Pricing and availability is not currently available.