- Suzuki-Miyaura cross-coupling reactions of benzyl halides with potassium aryltrifluoroborates.
Suzuki-Miyaura cross-coupling reactions of benzyl halides with potassium aryltrifluoroborates.
The Journal of organic chemistry (2006-11-18)
Gary A Molander, Maxwell D Elia
PMID17109547
ABSTRACT
[reaction: see text] The palladium-catalyzed cross-coupling of potassium aryltrifluoroborates with benzylic halides occurs in good yield with high functional group tolerance. The increased stability of potassium aryltrifluoroborates compared to other boron coupling partners makes this an effective route to functionalized methylene-linked biaryl systems.