Merck
  • Two distinct integrin-mediated mechanisms contribute to apical lumen formation in epithelial cells.
Two distinct integrin-mediated mechanisms contribute to apical lumen formation in epithelial cells.
PloS one (2011-05-17)
Satu Marja Myllymäki, Terhi Piritta Teräväinen, Aki Manninen
ABSTRACT
Formation of apical compartments underlies the morphogenesis of most epithelial organs during development. The extracellular matrix (ECM), particularly the basement membrane (BM), plays an important role in orienting the apico-basal polarity and thereby the positioning of apical lumens. Integrins have been recognized as essential mediators of matrix-derived polarity signals. The importance of β1-integrins in epithelial polarization is well established but the significance of the accompanying α-subunits have not been analyzed in detail. Here we demonstrate that two distinct integrin-dependent pathways regulate formation of apical lumens to ensure robust apical membrane biogenesis under different microenvironmental conditions; 1) α2β1- and α6β4-integrins were required to establish a basal cue that depends on Rac1-activity and guides apico-basal cell polarization. 2) α3β1-integrins were implicated in positioning of mitotic spindles in cysts, a process that is essential for Cdc42-driven epithelial hollowing. Identification of the separate processes driven by particular integrin receptors clarifies the functional hierarchies between the different integrins co-expressed in epithelial cells and provides valuable insight into the complexity of cell-ECM interactions thereby guiding future studies addressing the molecular basis of epithelial morphogenesis during development and disease.
Sigma-Aldrich
Laminin from human placenta, liquid, BioReagent, suitable for cell culture
Pricing and availability is not currently available.