Merck
  • Atopic dermatitis induces the expansion of thymus-derived regulatory T cells exhibiting a Th2-like phenotype in mice.
Atopic dermatitis induces the expansion of thymus-derived regulatory T cells exhibiting a Th2-like phenotype in mice.
Journal of cellular and molecular medicine (2016-03-05)
Verena Moosbrugger-Martinz, Christoph H Tripp, Björn E Clausen, Matthias Schmuth, Sandrine Dubrac
ABSTRACT
Atopic dermatitis (AD) is a widespread inflammatory skin disease with an early onset, characterized by pruritus, eczematous lesions and skin dryness. This chronic relapsing disease is believed to be primarily a result of a defective epidermal barrier function associated with genetic susceptibility, immune hyper-responsiveness of the skin and environmental factors. Although the important role of abnormal immune reactivity in the pathogenesis of AD is widely accepted, the role of regulatory T cells (Tregs) remains elusive. We found that the Treg population is expanded in a mouse model of AD, i.e. mice topically treated with vitamin D3 (VitD). Moreover, mice with AD-like symptoms exhibit increased inducible T-cell costimulator (ICOS)-, cytotoxic T-lymphocyte antigen-4 (CTLA-4)- and Glycoprotein-A repetitions predominant receptor (GARP)-expressing Tregs in skin-draining lymph nodes. Importantly, the differentiation of Tregs into thymus-derived Tregs is favoured in our mouse model of AD. Emigrated skin-derived dendritic cells are required for Treg induction and Langerhans cells are responsible for the biased expansion of thymus-derived Tregs . Intriguingly, thymus-derived Tregs isolated from mice with AD-like symptoms exhibit a Th2 cytokine profile. Thus, AD might favour the expansion of pathogenic Tregs able to produce Th2 cytokines and to promote the disease instead of alleviating symptoms.