Merck
  • Potential of ezetimibe in memory deficits associated with dementia of Alzheimer's type in mice.
Potential of ezetimibe in memory deficits associated with dementia of Alzheimer's type in mice.
Indian journal of pharmacology (2010-04-22)
Yogita Dalla, Nirmal Singh, Amteshwar Singh Jaggi, Dhandeep Singh, Pooja Ghulati
ABSTRACT
High cholesterol levels have been positively correlated with a higher incidence of memory impairment and dementia. The study was undertaken to investigate the potential of the lipid-lowering drug, ezetimibe, in memory deficits associated with dementia of Alzheimer's (AD) type in mice. Dementia was induced with chronic administration of a high-fat diet (HFD) or intracebroventricular streptozotocin (ICV STZ, two doses of 3 mg/kg) in separate groups of animals. The memory of the animals was assessed by employing a Morris water maze. Brain thio barbituric acid-reactive species and reduced glutathione levels were measured to assess the total oxidative stress. Brain acetyl cholinesterase (AChE) activity and total serum cholesterol levels were also measured. STZ/HFD produced a significant impairment of memory along with an increase in brain AChE activity and oxidative stress. HFD mice also showed an increase in cholesterol levels. Ezetimibe (10 mg/kg, orally for 15 days) significantly attenuated STZ/HFD-induced memory deficits and biochemical changes. It also prevented HFD-induced rise in the cholesterol level. The memory-restorative effect of ezetimibe can be attributed to its cholesterol-dependent as well as cholesterol-independent effects. The study highlights the potential of ezetimibe in memory dysfunctions associated with dementia of AD.
Sigma-Aldrich
1-Methoxypropane, 97%