Merck
Telmisartan: a review of its use in hypertension.
Drugs (2001-09-18)
M Sharpe, B Jarvis, K L Goa
ABSTRACT
Telmisartan is an angiotensin II receptor antagonist that is highly selective for type 1 angiotensin II receptors. It was significantly more effective than placebo in large (n >100), double-blind, randomised, multicentre clinical trials in patients with mild to moderate hypertension. Telmisartan 20 to 160 mg once daily produced mean reductions in supine trough systolic blood pressure and diastolic blood pressure of up to 15.5 and 10.5 mm Hg, respectively. Maximum blood pressure reduction occurred with a dosage of 40 to 80 mg/day. Telmisartan 40 to 120 mg/day was as effective as amlodipine 5 to 10 mg/day or atenolol 50 to 100 mg/day in dose-titration studies. Telmisartan 20 to 160 mg/day was generally similar in efficacy to enalapril 5 to 20 mg/day or lisinopril 10 to 40 mg/day in both titration-to-response and other studies. Hydrochlorothiazide was coadministered in most of the titration-to-response studies if patients remained hypertensive. Telmisartan 80 mg/day was more effective than submaximal dosages of losartan (50 mg/day) or valsartan (80 mg/day) and was as effective as a fixed-dose combination of losartan 50 mg plus hydrochlorothiazide 12.5 mg over the last 6 hours of the dosage interval and the whole 24-hour postdose interval. In patients with severe hypertension, telmisartan 80 to 160 mg/day was as effective as enalapril 20 to 40 mg/day (both agents could be titrated and combined sequentially with hydrochlorothiazide 25 mg and amlodipine 5 mg). The addition of hydrochlorothiazide to telmisartan was more effective than each agent alone at lowering blood pressure in patients with hypertension. Telmisartan was well tolerated in patients with mild to moderate hypertension and was significantly less likely to cause persistent, dry cough than lisinopril. Telmisartan is an effective antihypertensive agent with a tolerability profile similar to that of placebo. Comparative data have shown telmisartan to be as effective as other major classes of antihypertensive agents at lowering blood pressure. Compared with lisinopril, telmisartan is associated with a significantly lower incidence of dry, persistent cough. Therefore, telmisartan is a useful therapeutic option in the management of patients with hypertension.
Sigma-Aldrich
Telmisartan, ≥98% (HPLC), solid
Pricing and availability is not currently available.