Merck
  • Hydrogen-bond-promoted hetero-Diels-Alder reactions of unactivated ketones.
Hydrogen-bond-promoted hetero-Diels-Alder reactions of unactivated ketones.
Journal of the American Chemical Society (2002-08-15)
Yong Huang, Viresh H Rawal
ABSTRACT
We report the first examples of hydrogen-bond-promoted acceleration of hetero-Diels-Alder reactions and the use of such catalysis for the hetero-Diels-Alder reactions of simple, unactivated ketones. Several spiro-fused dihydropyans were synthesized in good yields using this procedure. This activation protocol represents an attractive and operationally simple alternative to conventional, Lewis acid catalysis.
Sigma-Aldrich
1-Z-4-Piperidone, 99%
Pricing and availability is not currently available.