Merck
  • A facile synthesis of radioiodinated (Z)-vinyl iodides via vinylboronates.
A facile synthesis of radioiodinated (Z)-vinyl iodides via vinylboronates.
Nuclear medicine and biology (2003-05-28)
George W Kabalka, Murthy R Akula, Jinhua Zhang
ABSTRACT
A direct radioiodination of (Z)-vinylboronic acid esters to the corresponding vinyl iodides using Na(123)I and chloramine-T is described. The boronates were prepared from vinyl iodides via palladium coupling reactions.
Sigma-Aldrich
Bis(neopentyl glycolato)diboron, 96%
Pricing and availability is not currently available.