Merck
Capillary zone electrophoresis of linear and branched oligosaccharides.
Journal of chromatography (1992-05-29)
W Nashabeh, Z el Rassi
ABSTRACT
The electrophoretic behavior of derivatized linear and branched oligosaccharides from various sources was examined in capillary zone electrophoresis with polyether-coated fused-silica capillaries. Two UV-absorbing (also fluorescent) derivatizing agents (2-aminopyridine and 6-aminoquinoline) were utilized for the electrophoresis and sensitive detection of neutral oligosaccharides, e.g., N-acetylchitooligosaccharides, high-mannose glycans and xyloglucan oligosaccharides. The oligosaccharides labelled with 6-aminoquinoline yielded eight times higher signal than those tagged with 2-aminopyridine. Plots of logarithmic electrophoretic mobilities of labelled N-acetylchitooligosaccharides with 6-aminoquinoline or 2-aminopyridine versus the number of sugar residues in the homologous series yielded straight lines in the size range studied, the slopes of which were independent of the tagging functions. The slopes of these lines are referred to as the N-acetylglucosaminyl group mobility decrement. The oligosaccharides were better resolved in the presence of tetrabutylammonium bromide in the running electrolyte. Furthermore, the electrophoretic mobilities of branched oligosaccharides were indexed with respect to linear homooligosaccharides, an approach that may prove valuable in correlating and predicting the mobilities of complex oligosaccharides.
Sigma-Aldrich
6-Aminoquinoline, 98%
Pricing and availability is not currently available.