Merck
New triazine derivatives as potent modulators of multidrug resistance.
Journal of medicinal chemistry (1992-06-26)
A Dhainaut, G Régnier, G Atassi, A Pierré, S Léonce, L Kraus-Berthier, J F Prost
ABSTRACT
A series of 70 triazine derivatives have been synthesized and tested for their capacity to modulate multidrug resistance (MDR) in DC-3F/AD and KB-A1 tumor cells in vitro, in comparison with verapamil (VRP), a calcium channel antagonist currently used in therapy as an antihypertensive drug, which also shows MDR modulating activity. Among the 12 selected compounds, 16 (S9788) showed high MDR reversing properties in vitro (300- and 6-fold VRP at 5 microM in DC-3F/AD and KB-A1 cells, respectively) and induced a strong accumulation of adriamycin. The relationship between the increase of ADR accumulation and the fold reversal induced by these compounds and their lack of effects on the sensitive DC-3F cells suggest that they act mainly by inhibiting the P-glycoprotein (Pgp) catalyzed efflux of cytotoxic agents, as already described for a majority of MDR modulators. In vivo, in association with the antitumor drug vincristine (0.25 mg/kg), 16 (100 mg/kg) increased the T/C by 39% in mice bearing the resistant tumor cell line P388/VCR. According to these interesting properties, 16 was selected for a clinical development because it was more bioavailable than 34, even though it was less active.
Sigma-Aldrich
5-Chlorodibenzosuberane, 96%