Merck
  • Synthesis and in vitro and in vivo antitumor activity of 2-phenylpyrroloquinolin-4-ones.
Synthesis and in vitro and in vivo antitumor activity of 2-phenylpyrroloquinolin-4-ones.
Journal of medicinal chemistry (2005-04-29)
Maria Grazia Ferlin, Gianfranco Chiarelotto, Venusia Gasparotto, Lisa Dalla Via, Vincenzo Pezzi, Luisa Barzon, Giorgio Palù, Ignazio Castagliuolo
ABSTRACT
In our search for potential new anticancer drugs, we designed and synthesized a series of tricyclic compounds containing the antimitotic 2-phenylazaflavone chromophore fused to a pyrrole ring in a pyrroloquinoline structure. Compounds 8, 18, 19, 22, 23, 25 and 26, when tested against a panel of fourteen human tumor cell lines, showed poor in vitro cytotoxic activity, whereas 20, 21 and 24 showed significant activity (IC(50) 0.7 to 50 microM). Steroid hormone-sensitive ovary, liver, breast and adrenal gland adenocarcinoma cell lines displayed the highest sensitivity (IC(50) 0.7 to 8 microM). Compound 24 blocked cells in the G(2)/M phase of the cell cycle and induced a significant increase in apoptotis. Compounds 20, 21 and 24 proved to alter microtubule assembly and stability, displaying a cytoplasmic microtubule network similar to that caused by Vincristine. In vivo, administration of compound 24 to Balb/c mice inhibited the growth of a syngenic hepatocellular carcinoma.
Sigma-Aldrich
4-Aminoindole, 97%
Pricing and availability is not currently available.