Merck
  • Large-scale production of liposomes containing monomeric zinc phthalocyanine by controlled dilution of organic solvents.
Large-scale production of liposomes containing monomeric zinc phthalocyanine by controlled dilution of organic solvents.
Journal of pharmaceutical sciences (1994-11-01)
U Isele, P van Hoogevest, R Hilfiker, H G Capraro, K Schieweck, H Leuenberger
ABSTRACT
This work describes the development of an organic solvent dilution method suitable for the large scale manufacturing of small unilamellar liposomes containing the water-insoluble photosensitizer zinc phthalocyanine in the monomeric state. N-Methyl pyrrolidone (NMP)/tert-butyl alcohol was selected as water miscible organic phase in which the phospholipids 1-palmitoyl, 2-oleoylphosphatidylcholine (POPC), and 1,2-dioeloylphosphatidylserine (OOPS) and the dye were dissolved. This organic phase was mixed with an excess of a water phase using a dynamic mixing device, yielding reproducibly unilamellar liposomes with a mean size of 50-150 nm as measured with quasielastic light scattering. After concentration, the organic solvents were efficiently removed by cross-flow filtration. The liposomes were then sterile filtered and freeze-dried. A method to measure the aggregation state of the dye in the liposomes was developed. A stable lyophilized formulation containing monomeric liposomal ZnPc could be obtained by using a solution of ZnPc in NMP (2 mg/mL) and ZnPc/phospholipid (1:100 w/w ratio) and performing concentration and dialysis at 4 degrees C and lyophilization in presence of a mixture of lactose and phospholipid (5:1 w/w ratio).
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Pricing and availability is not currently available.