Merck
  • Synthesis and Crystal Structures of Benzohydroxamate-Coordinated Vanadium(V) Oxo Complexes with Aroylhydrazone Ligands.
Synthesis and Crystal Structures of Benzohydroxamate-Coordinated Vanadium(V) Oxo Complexes with Aroylhydrazone Ligands.
Acta chimica Slovenica (2015-12-19)
Shao-Song Qian, Xinlu Zhao, Jia Wang, Zhonglu You
ABSTRACT
Reaction of [VO(acac)(2)] (where acac = acetylacetonate), benzohydroxamic acid (Hbha), and two similar aroylhydrazone ligands in methanol produced two benzohydroxamate-coordinated mononuclear vanadium(V) oxo complexes with general formula [VOL(bha)], where L = L(1) = N'-(5-bromo-2-hydroxybenzylidene)-2-fluorobenzohydrazide (H(2)L(1)), and L = L(2) = N'-(3-bromo-2-hydroxybenzylidene)-2-fluorobenzohydrazide (H(2)L(2)). Crystal and molecular structures of the complexes were determined by single crystal X-ray diffraction method. All of the investigated compounds were further characterized by elemental analysis, and FT-IR and UV-Vis spectra. Single crystal X-ray structural studies indicate that the benzohydrazone ligands coordinate to the VOcores through phenolate O, imino N, and enolate O atoms, and the benzohydroxamate ligands coordinate to the VO cores through deprotonated hydroxyl O and carbonyl O atoms. The V atoms in both complexes are in octahedral coordination. Thermal stability of the complexes was studied.
Sigma-Aldrich
2-Fluorobenzoic hydrazide, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
5-Bromosalicylaldehyde, 98%
Pricing and availability is not currently available.