Merck
  • Occurrence and Profiles of the Artificial Endocrine Disruptor Bisphenol A and Natural Endocrine Disruptor Phytoestrogens in Urine from Children in China.
Occurrence and Profiles of the Artificial Endocrine Disruptor Bisphenol A and Natural Endocrine Disruptor Phytoestrogens in Urine from Children in China.
International journal of environmental research and public health (2015-12-04)
Mingyue Zhang, Zhenghua Duan, Yinghong Wu, Zhen Liu, Ke Li, Lei Wang
ABSTRACT
Exposure to artificial or natural endocrine disruptors, such as bisphenol A (BPA) and phytoestrogens has been demonstrated to have health effects, especially in children. Biomonitoring of BPA and phytoestrogens in human urine can be used to assess the intake levels of these compounds. In this study, BPA and phytoestrogens in urine specimens (n = 256) collected from children in China were measured by liquid chromatography (LC)-tandem mass spectrometry (MS/MS). BPA was detected in most specimens, with a geometric mean concentration of 1.58 ng/mL. For the first time, levels of urinary phytoestrogens in Chinese children were reported. Daidzein and enterolactone are the typical isoflavones and lignans compounds in urine, respectively. Relatively high levels of urinary BPA indicate an increasing risk of BPA exposure to Chinese children. Urinary concentrations of daidzein in Chinese children are higher when compared with those reported in the U.S. children, while concentrations of urinary enterolactone and enterodiols are significantly lower. This suggests a significant difference in phytoestrogen intake between the children from China and from the U.S.
Sigma-Aldrich
Enterodiol, ≥95.0% (HPLC)
Sigma-Aldrich
Enterolactone, ~95% (HPLC)