Merck
  • Hydrothermal synthesis, structure and magnetic properties of a three-dimensional cobalt(II) - aminophenyltetrazolate coordination polymer.
Hydrothermal synthesis, structure and magnetic properties of a three-dimensional cobalt(II) - aminophenyltetrazolate coordination polymer.
Dalton transactions (Cambridge, England : 2003) (2014-04-01)
Tiffany M Smith, Michael Tichenor, Yuan-Zhu Zhang, Kim R Dunbar, Jon Zubieta
ABSTRACT
The hydrothermal reaction of cobalt(II) acetate tetrahydrate, 4-aminophenyl tetrazole and water at 135 °C for 48 hours provided orange crystals of [Co3(OH)2(H2O)2(aptet)4] (1) (aptet = 4-aminophenyltetrazolate). The three-dimensional structure of compound 1 is constructed from chains of corner- and edge-sharing Co(II) octahedra linked into a framework through the dipodal ligand tethers. The cobalt sites of the chain are linked through tetrazolate groups in the N2,N3-bridging mode, as well as through μ3-hydroxyl groups into trinuclear Co3 substructures. The magnetic properties are consistent with a ferrimagnetic chain with the non-compensating resultant moment of one Co(II) per trinuclear Co(II) subunit. The temperature dependence of the ac magnetic susceptibility indicates glassy-like magnetic behavior.
Sigma-Aldrich
Cobalt(II) acetate tetrahydrate, reagent grade
Pricing and availability is not currently available.
Sigma-Aldrich
Cobalt(II) acetate tetrahydrate, ACS reagent, ≥98.0%
Pricing and availability is not currently available.