Merck
  • Potassium iodate assisted synthesis of titanium dioxide nanoparticles with superior water-dispersibility.
Potassium iodate assisted synthesis of titanium dioxide nanoparticles with superior water-dispersibility.
Journal of colloid and interface science (2014-07-07)
Yawen Wang, Fangfang Duo, Shiqi Peng, Falong Jia, Caimei Fan
ABSTRACT
In this paper, we report a novel polyol process to synthesize highly water-dispersible anatase titanium dioxide (TiO2) nanoparticles (∼5 nm) by the introduction of inorganic oxidizing agent--KIO3. The obtained TiO2 nanoparticles are well dispersible in water at pH≥5.0 and the resulting aqueous dispersion remains stable over months. The superior water-dispersibility of as-formed TiO2 is ascribed to the electrostatic repulsion from carboxylic acid group modified on TiO2 nanoparticles, which is the oxidation product of solvent diethylene glycol (DEG) by KIO3. Based on the characterization results, the formation processes of water-dispersibility TiO2 nanoparticles are proposed. Meanwhile, the synthesized TiO2 nanoparticles are found to be doped by iodine and exhibit excellent photocatalytic activity on degradation of rhodamine-B (RhB) under visible-light irradiation. The further tests demonstrate that the O(2-) is the main active species during photodegradation of RhB.
Sigma-Aldrich
Potassium iodate, reagent grade, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium iodate, ACS reagent, 99.5%
Pricing and availability is not currently available.