Merck
  • Silencing of MICAL-L2 suppresses malignancy of ovarian cancer by inducing mesenchymal-epithelial transition.
Silencing of MICAL-L2 suppresses malignancy of ovarian cancer by inducing mesenchymal-epithelial transition.
Cancer letters (2015-04-14)
Lin-Yan Zhu, Wen-Ming Zhang, Xiao-Mei Yang, Lining Cui, Jun Li, Yan-Li Zhang, Ya-Hui Wang, Jun-Ping Ao, Ming-Ze Ma, Huan Lu, Yuan Ren, Shao-Hua Xu, Guang-Dong Yang, Wei-Wei Song, Jing-Hao Wang, Xiao-Dan Zhang, Rong Zhang, Zhi-Gang Zhang
ABSTRACT
Ovarian cancer remains the disease with the highest associated mortality rate of gynecologic malignancy due to cancer metastasis. Rearrangement of actin cytoskeleton by cytoskeleton protein plays a critical role in tumor cell metastasis. MICAL-L2, a member of MICAL family, can interact with actin-binding proteins, regulate actin cross-linking and coordinate the assembly of adherens junctions and tight junctions. However, the roles of MICAL-L2 in tumors and diseases have not been explored. In this study, we found that MICAL-L2 protein is significantly up-regulated in ovarian cancer tissues along with FIGO stage and associated with histologic subgroups of ovarian cancer. Silencing of MICAL-L2 suppressed ovarian cancer cell proliferation, migration and invasion ability. Moreover, silencing of MICAL-L2 prevented nuclear translocation of β-catenin, inhibited canonical wnt/β-catenin signaling and induced the mesenchymal-epithelial transition (MET). Taken together, our data indicated that MICAL-L2 may be an important regulator of epithelial-mesenchymal transition (EMT) in ovarian cancer cells and a new therapeutic target for interventions against ovarian cancer invasion and metastasis.