Merck
  • Behavior of Multiclass Pesticide Residue Concentrations during the Transformation from Rose Petals to Rose Absolute.
Behavior of Multiclass Pesticide Residue Concentrations during the Transformation from Rose Petals to Rose Absolute.
Journal of agricultural and food chemistry (2015-05-06)
Oriane Tascone, Yoann Fillâtre, Céline Roy, Uwe J Meierhenrich
ABSTRACT
This study investigates the concentrations of 54 multiclass pesticides during the transformation processes from rose petal to concrete and absolute using roses spiked with pesticides as a model. The concentrations of the pesticides were followed during the process of transforming the spiked rose flowers from an organic field into concrete and then into absolute. The rose flowers, the concrete, and the absolute, as well as their transformation intermediates, were analyzed for pesticide content using gas chromatography/tandem mass spectrometry. We observed that all the pesticides were extracted and concentrated in the absolute, with the exception of three molecules: fenthion, fenamiphos, and phorate. Typical pesticides were found to be concentrated by a factor of 100-300 from the rose flowers to the rose absolute. The observed effect of pesticide enrichment was also studied in roses and their extracts from four classically phytosanitary treated fields. Seventeen pesticides were detected in at least one of the extracts. Like the case for the spiked samples in our model, the pesticides present in the rose flowers from Turkey were concentrated in the absolute. Two pesticides, methidathion and chlorpyrifos, were quantified in the rose flowers at approximately 0.01 and 0.01-0.05 mg kg(-1), respectively, depending on the treated field. The concentrations determined for the corresponding rose absolutes were 4.7 mg kg(-1) for methidathion and 0.65-27.25 mg kg(-1) for chlorpyrifos.
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
Methane, electronic grade, ≥99.998%
Sigma-Aldrich
Citronellyl acetate, ≥95%, FCC, FG
Sigma-Aldrich
Carbon nanofibers, graphitized (iron-free), composed of conical platelets, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Geranyl acetate, natural, FCC
Sigma-Aldrich
β-Citronellol, 95%
Sigma-Aldrich
Geranyl acetate, ≥97%
Sigma-Aldrich
2-Phenylethanol, ≥99.0% (GC)
Sigma-Aldrich
Phenethyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Geranyl acetate, FCC
Sigma-Aldrich
Phenethyl alcohol, ≥99%, FCC, FG