Merck
  • Possible functional groups responsible for inhibition of in vivo angiogenesis by herbimycin A.
Possible functional groups responsible for inhibition of in vivo angiogenesis by herbimycin A.
Biological & pharmaceutical bulletin (1994-10-01)
T Oikawa, H Ogasawara, H Sano, K Shibata, S Omura
ABSTRACT
Six herbimycin A (HBM) derivatives were examined for their anti-angiogenic effects in a bioassay system involving chorioallantoic membranes (CAMs) of growing chick embryos on the basis of our previous observation that HBM is a potent angiogenesis inhibitor. 17-Cyclopropylamino-HBM dose-dependently inhibited embryonic angiogenesis. The ID50 value was 0.1 microgram (160 pmol) per egg and thereby lower than that of the parent compound HBM (ID50 = 0.15 micrograms (260 pmol) per egg). In contrast, 19-dimethylamino-, N-acetyl-, 2,3,4,5-tetrahydro- and 7-decarbamoyl-HBM at doses of 0.01-10 micrograms/egg failed to affect angiogenesis in CAMs. These results strongly suggest as follows: (1) C-19 position, amino group between positions C-1 and C-20 and carbamoyl group in C-7 are essential for the anti-angiogenic action of HBM; (2) HBM needs certain fixed conformation for expression of angiogenesis inhibition; (3) it is expected that the modification of C-17 with a suitable functional group results in increased anti-angiogenic potency of HBM--that is, a more potent angiogenesis inhibitor than the parent compound would be developed.
Sigma-Aldrich
Herbimycin A from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Pricing and availability is not currently available.