Merck
  • Coronary vasospasm induced in transgenic mouse with increased phospholipase C-δ1 activity.
Coronary vasospasm induced in transgenic mouse with increased phospholipase C-δ1 activity.
Circulation (2012-01-24)
Shuji Shibutani, Tomohiro Osanai, Toshihiro Ashitate, Shigeki Sagara, Kei Izumiyama, Yuko Yamamoto, Kenji Hanada, Takashi Echizen, Hirofumi Tomita, Takeshi Fujita, Takeshi Miwa, Hiroaki Matsubara, Yoshimi Homma, Ken Okumura
ABSTRACT
We reported that phospholipase C (PLC)-δ1 activity was enhanced 3-fold in patients with coronary spastic angina. We detected variant PLC-δ1 with replacement of arginine 257 by histidine (R257H) showing increased enzymatic activity. We tested the hypothesis that increased PLC-δ1 activity causes enhanced coronary vasomotility. We generated transgenic (TG) mice with human R257H variant PLC-δ1 in vascular smooth muscle cells. PLC enzymatic activity in the coronary artery was increased by 2.57 and 1.89 times, respectively, in homozygous and heterozygous TG compared with wild-type (WT) mice. ST elevation after ergometrine occurred in 17 of 18 homozygous TG, 6 of 20 heterozygous TG, and 3 of 22 WT mice (P<0.01, homozygous TG versus WT; P<0.05, homozygous TG versus heterozygous TG; P=NS, heterozygous TG versus WT). ST elevation was associated with bradyarrhythmias in homozygous TG mice. Focal coronary artery narrowing was documented with the microvascular filling technique in 3 of 5 homozygous TG mice after ergometrine but not in any of 7 WT mice (P<0.05). In the isolated Langendorff hearts, coronary perfusion pressure was increased after ergometrine in homozygous TG mice (P<0.01) but not in heterozygous TG or WT mice. Coronary perfusion pressure increase after prostaglandin F2α was similar among homozygous TG, heterozygous TG, and WT mice. Cultured rat aortic smooth muscle cells transfected with variant PLC-δ1 showed a higher PLC activity than those with WT PLC-δ1 (P<0.05) and furthermore showed greater intracellular Ca2+ response to acetylcholine in variant than in WT PLC-δ1 (P<0.05). Increased PLC-δ1 activity enhances coronary vasomotility such as that seen in patients with coronary spastic angina.