Merck
  • Does minor histocompatibility antigen HA-1 disparity affect the occurrence of graft-versus-host disease in tunisian recipients of hematopoietic stem cells?
Does minor histocompatibility antigen HA-1 disparity affect the occurrence of graft-versus-host disease in tunisian recipients of hematopoietic stem cells?
Clinics (Sao Paulo, Brazil) (2011-01-19)
Mohamed Hichem Sellami, Lamia Torjemane, Alejandro Espadas de Arias, Houda Kaabi, Saloua Ladeb, Francesca Poli, Tarek Ben Othmane, Slama Hmida
ABSTRACT
Minor histocompatibility antigen HA-1 (MiHAg-HA-1) disparity between a patient and his or her human leukocyte antigen (HLA) genoidentical donor has been widely associated with an increased risk of graft-versus-host disease following allogeneic hematopoietic stem cell transplantation. To examine the effect of HA-1 disparity on the incidence of both acute and chronic graft-versus-host disease in Tunisian recipients of hematopoietic stem cells. A total of 60 patients and their 60 respective sibling hematopoietic stem cell donors were enrolled in this study. All patients prophylactically received cyclosporine A and/or methotrexate for graft-versus-host disease. An HA-1 genotyping assay was performed with the SSP-PCR method, and HLA-A*0201- and/or HLA-A*0206-positive samples were identified using the Luminex HLA typing method. The Luminex HLA typing assay showed that 54 patients were positive for either the HLA-A*0201 or HLA-A*0206 alleles. Among these cases, six pairs were mismatched for MiHAg-HA-1. Both acute and chronic graft-versus-host disease occurred in four mismatched patients (Fisher's p-values were 0.044 and 0.170, respectively). A univariate logistic regression model analysis showed that only acute graft-versus-host disease may be affected by recipient MiHAg-HA-1 disparity (p: 0.041, OR: 6.727), while chronic graft-versus-host disease correlates with both age and recipient/donor sex mismatch (p: 0.014, OR: 8.556 and p: 0.033, OR: 8.664, respectively). Our findings support previously reported data suggesting a significant association between HA-1 disparity and the risk of acute graft-versus-host disease following hematopoietic stem cell transplantation.