Merck
Terconazole - a new broad-spectrum antifungal.
Chemotherapy (1983-01-01)
J Van Cutsem, F Van Gerven, R Zaman, P A Janssen
ABSTRACT
Terconazole, a new triazole ketal, is found to be highly active in vitro on a wide range of yeasts and mycelium-forming fungi. The in vitro activity depends largely on the medium used. In vitro it is a potent antifungal agent in preventing the morphogenetic transformation of the yeast into the (pseudo-)mycelium form of Candida albicans. In vivo terconazole is highly active in topical treatment of various experimental models of dermatophytosis and candidosis. It also possesses moderate oral broad-spectrum activity. No side effects were observed.
Sigma-Aldrich
Terconazole, ≥98%