Merck
  • Expression of fibroblast growth factor 9 in normal human lung and idiopathic pulmonary fibrosis.
Expression of fibroblast growth factor 9 in normal human lung and idiopathic pulmonary fibrosis.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2013-06-26)
Emily Coffey, Donna R Newman, Philip L Sannes
ABSTRACT
The fibroblast growth factor (FGF) family of signaling ligands contributes significantly to lung development and maintenance in the adult. FGF9 is involved in control of epithelial branching and mesenchymal proliferation and expansion in developing lungs. However, its activity and expression in the normal adult lung and by epithelial and interstitial cells in fibroproliferative diseases like idiopathic pulmonary fibrosis (IPF) are unknown. Tissue samples from normal organ donor human lungs and those of a cohort of patients with mild to severe IPF were sectioned and stained for the immunolocalization of FGF9. In normal lungs, FGF9 was confined to smooth muscle surrounding airways, alveolar ducts and sacs, and blood vessels. In addition to these same sites, lungs of IPF patients expressed FGF9 in a population of myofibroblasts within fibroblastic foci, hypertrophic and hyperplastic epithelium of airways and alveoli, and smooth muscle cells surrounding vessels embedded in thickened interstitium. The results demonstrate that FGF9 protein increased in regions of active cellular hyperplasia, metaplasia, and fibrotic expansion of IPF lungs, and in isolated human lung fibroblasts treated with TGF-β1 and/or overexpressing Wnt7B. The cellular distribution and established biologic activity of FGF9 make it a potentially strong candidate for contributing to the progression of IPF.
Sigma-Aldrich
Fibroblast Growth Factor-9 human, recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)
Pricing and availability is not currently available.
Sigma-Aldrich
FGF-9 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Pricing and availability is not currently available.