Merck
  • The p21-activated kinase PAK is negatively regulated by POPX1 and POPX2, a pair of serine/threonine phosphatases of the PP2C family.
The p21-activated kinase PAK is negatively regulated by POPX1 and POPX2, a pair of serine/threonine phosphatases of the PP2C family.
Current biology : CB (2002-02-28)
Cheng-Gee Koh, E-Jean Tan, Edward Manser, Louis Lim
ABSTRACT
The Rho GTPases are involved in many signaling pathways and cellular functions, including the organization of the actin cytoskeleton, regulation of transcription, cell motility, and cell division. The p21 (Cdc42/Rac)-activated kinase PAK mediates a number of biological effects downstream of these Rho GTPases (reviewed by [1]). The phosphorylation state of mammalian PAK is highly regulated: upon binding of GTPases, PAK is potently activated by autophosphorylation at multiple sites, although the mechanisms of PAK downregulation are not known. We now report two PP2C-like serine/threonine phosphatases (POPX1 and POPX2) that efficiently inactivate PAK. POPX1 was isolated as a binding partner for the PAK interacting guanine nucleotide exchange factor PIX. The dephosphorylating activity of POPX correlates with an ability to block the in vivo effects of active PAK. Consonant with these effects on PAK, POPX can also inhibit actin stress fiber breakdown and morphological changes driven by active Cdc42(V12). The association of the POPX phosphatases with PAK complexes may allow PAK to cycle rapidly between active and inactive states; it represents a unique regulatory component of the signaling pathways of the PAK kinase family.