Merck
  • Association between serum alpha-fetoprotein levels and fatty liver disease: a cross-sectional study.
Association between serum alpha-fetoprotein levels and fatty liver disease: a cross-sectional study.
World journal of gastroenterology (2014-09-11)
Ping Xu, Cheng-Fu Xu, Xing-Yong Wan, Chao-Hui Yu, Chao Shen, Peng Chen, Gen-Yun Xu, You-Ming Li
ABSTRACT
To investigate the association between serum alpha-fetoprotein (AFP) levels and fatty liver disease (FLD) in a Chinese population. A cross-sectional study was performed among subjects who presented for a health examination at the First Affiliated Hospital, College of Medicine, Zhejiang University in 2013. FLD was diagnosed based on an ultrasonography examination. Serum AFP levels were measured with a chemiluminescence immunoassay. Of the 9800 subjects enrolled, 2601 were diagnosed with FLD. Subjects with FLD had higher serum AFP levels than those without the disease. Subjects with high serum AFP levels had a higher prevalence of FLD, metabolic syndrome, and its components. Univariate logistic analysis showed that elevated serum AFP levels were associated with an increased risk of FLD (OR = 1.057, 95%CI: 1.031-1.084). However, after adjusting for covariates, AFP no longer remained significantly associated with the risk factors for FLD. Our results suggest that serum AFP levels are significantly associated with FLD and that AFP acts as a cofactor, but not as an independent factor, for FLD.
Sigma-Aldrich
Human AFP / Alpha-fetoprotein ELISA Kit, for serum, plasma, cell culture supernatants and urine
Pricing and availability is not currently available.