Merck
  • Dictyosphaeric acids A and B: new decalactones from an undescribed Penicillium sp. obtained from the alga Dictyosphaeria versluyii.
Dictyosphaeric acids A and B: new decalactones from an undescribed Penicillium sp. obtained from the alga Dictyosphaeria versluyii.
Journal of natural products (2004-08-31)
Tim S Bugni, Jeffrey E Janso, R Thomas Williamson, Xidong Feng, Valerie S Bernan, Michael Greenstein, Guy T Carter, William M Maiese, Chris M Ireland
ABSTRACT
Fungal isolate F01V25 was obtained from the alga Dictyosphaeria versluyii collected near Dravuni, Fiji, in 2001 and represented a previously undescribed Penicillium sp. Fermentation of isolate F01V25 resulted in the production of two new polyketides, dictyosphaeric acids A and B, along with the known anthraquinone carviolin. The relative stereochemistry of dictyosphaeric acids A and B was determined using the J-based configuration analysis method in conjunction with ROE and NOE correlations.