Merck
  • Structure-activity relationship studies of 4-benzyl-1H-pyrazol-3-yl β-d-glucopyranoside derivatives as potent and selective sodium glucose co-transporter 1 (SGLT1) inhibitors with therapeutic activity on postprandial hyperglycemia.
Structure-activity relationship studies of 4-benzyl-1H-pyrazol-3-yl β-d-glucopyranoside derivatives as potent and selective sodium glucose co-transporter 1 (SGLT1) inhibitors with therapeutic activity on postprandial hyperglycemia.
Bioorganic & medicinal chemistry (2012-10-16)
Nobuhiko Fushimi, Hideki Fujikura, Hiroaki Shiohara, Hirotaka Teranishi, Kazuo Shimizu, Shigeru Yonekubo, Kohsuke Ohno, Takashi Miyagi, Fumiaki Itoh, Toshihide Shibazaki, Masaki Tomae, Yukiko Ishikawa-Takemura, Takeshi Nakabayashi, Noboru Kamada, Tomonaga Ozawa, Susumu Kobayashi, Masayuki Isaji
ABSTRACT
Sodium glucose co-transporter 1 (SGLT1) plays a dominant role in the absorption of glucose in the gut and is considered a promising target in the development of treatments for postprandial hyperglycemia. A series of 4-benzyl-1H-pyrazol-3-yl β-d-glucopyranoside derivatives have been synthesized, and its inhibitory activity toward SGLTs has been evaluated. By altering the substitution groups at the 5-position of the pyrazole ring, and every position of the phenyl ring, we studied the structure-activity relationship (SAR) profiles and identified a series of potent and selective SGLT1 inhibitors. Representative derivatives showed a dose-dependent suppressing effect on the escalation of blood glucose levels in oral mixed carbohydrate tolerance tests (OCTT) in streptozotocin-nicotinamide-induced diabetic rats (NA-STZ rats).
Sigma-Aldrich
Benzyl glucopyranoside, ≥95% (LC/MS-ELSD)