Merck
  • Effects of nanoparticle size and gestational age on maternal biodistribution and toxicity of gold nanoparticles in pregnant mice.
Effects of nanoparticle size and gestational age on maternal biodistribution and toxicity of gold nanoparticles in pregnant mice.
Toxicology letters (2014-08-08)
Hui Yang, Libo Du, Xin Tian, Zhenlin Fan, Cuiji Sun, Yang Liu, Jeffrey A Keelan, Guangjun Nie
ABSTRACT
Gold nanoparticles (GNPs) have considerable applications in biomedicine, such as in bio-sensing, bio-imaging, drug delivery and photothermal therapeutics. However, currently there are limited information regarding the impact of pregnancy on their biodistribution, elimination and toxicity. In this study, we investigated the biodistribution and potential toxic effects of different-sized GNPs (1.5, 4.5, 13, 30 and 70 nm in diameter) in non-pregnant and pregnant mice at different gestational ages (E5.5, 7.5, 9.5, 11.5 and 13.5). 5h after intravenous injection, GNPs exhibited size-dependent biodistribution profiles; however, regardless of size, no significant biodistribution changes were observed between non-pregnant and pregnant mice. Kinetic studies showed that 4.5 nm GNPs were primarily excreted through urine within 5h, whereas 30 nm GNPs had a more prolonged blood circulation time. No apparent toxic effects (e.g., increased mortality, altered behavior, reduced animal weight, abnormal organ morphology or reduced pregnancy duration) were observed with different-sized GNPs in pregnant mice. However, treatment with 30 nm GNPs induced mild emphysema-like changes in lungs of pregnant mice. These results indicated that the maternal biodistribution patterns of GNPs in pregnant mice depended on particle size, but not gestational age; organ-specific adverse effects may arise with treatment with some GNPs according to their size.
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3.2 mmol/g loading
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3 mmol/g triphenylphosphine loading
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3.0 mmol/g loading, 2 % cross-linked with divinylbenzene
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)