Merck
  • The IL-17A-producing CD8+ T-cell population in psoriatic lesional skin comprises mucosa-associated invariant T cells and conventional T cells.
The IL-17A-producing CD8+ T-cell population in psoriatic lesional skin comprises mucosa-associated invariant T cells and conventional T cells.
The Journal of investigative dermatology (2014-06-20)
Marcel B M Teunissen, Nataliya G Yeremenko, Dominique L P Baeten, Saskia Chielie, Phyllis I Spuls, Menno A de Rie, Olivier Lantz, Pieter C M Res
ABSTRACT
IL-17A is pivotal in the etiology of psoriasis, and CD8(+) T cells with the ability to produce this cytokine (Tc17 cells) are over-represented in psoriatic lesions. Here we demonstrate that the frequency of Tc17 cells in peripheral blood of psoriasis patients correlated with the clinical severity of the disease. Analysis of cutaneous-associated lymphocyte antigen expression showed that the blood Tc17 population contains a significantly higher proportion of cells with skin-homing potential compared with the CD8(+) T-cell population lacking IL-17A/IL-22 expression. IL-17A-producing CD8(+) T cells in blood have previously been reported to belong mainly to the mucosa-associated invariant T-cell (MAIT cell) lineage characterized by TCR Vα7.2 chain, CD161, IL-18Rα, and multidrug transporter ABCB1 expression. We demonstrate the presence of CD8(+) MAIT cells in the dermis and epidermis of psoriatic plaques, as well as healthy skin; however, IL-17A-producing CD8(+) MAIT cells were predominantly found in psoriatic skin. Notably, we observed IL-17A production in a large proportion of psoriatic plaque-derived CD8(+) T cells devoid of MAIT cell characteristics, likely representing conventional CD8(+) T cells. In conclusion, we provide supporting evidence that implicates Tc17 cells in the pathogenesis of psoriasis and describe the presence of innate CD8(+) MAIT cells in psoriatic lesions as an alternative source of IL-17A.
Sigma-Aldrich
(±)-CPP, solid
Pricing and availability is not currently available.
Ciclosporin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Ciclosporin for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.