Merck
  • A dual tyrosine kinase inhibitor lapatinib suppresses overexpression of matrix metallopeptidase 1 (MMP1) in endometrial cancer.
A dual tyrosine kinase inhibitor lapatinib suppresses overexpression of matrix metallopeptidase 1 (MMP1) in endometrial cancer.
Journal of molecular medicine (Berlin, Germany) (2014-05-14)
Chiao-Yun Lin, Angel Chao, Tzu-Hao Wang, Swei Hsueh, Yun-Shien Lee, Tzu-I Wu, An-Shine Chao, Huei-Jean Huang, Hung-Hsueh Chou, Ting-Chang Chang, Chyong-Huey Lai
ABSTRACT
Endometrial cancers have been recently molecularly characterized; amplifications of human epidermal growth factor receptor 2 (HER2) were seen in 25 % of the serous-like tumors, and mutations in the PI(3)K/AKT pathways were seen in 93 % of endometrioid tumors. These new findings about endometrial cancer suggest a potential for targeted therapy with lapatinib, a dual inhibitor of epidermal growth factor receptor and HER2 tyrosine kinases. However, the clinical efficacy of lapatinib in phase II clinical trials for the treatment of endometrial cancers was only minimal. In this study, we investigated the signaling changes induced by lapatinib in endometrial cancer, which may improve its therapeutic efficacy in molecularly selected patient groups. We identified one of the final molecular targets of lapatinib to be interstitial collagenase, matrix metallopeptidase 1 (MMP1). Lapatinib suppresses MMP1 through EGFR and HER2, and their downstream ERK and AKT signaling pathways. We also found that the activating protein-1 binding site of MMP1 promoter is required for its transcriptional activation, which may be unique for endometrial cancers. Our results also showed that forced expression of active ERK or active AKT mutants rescued MMP1 expression from lapatinib suppression, further suggesting the importance of molecular selection to find appropriate patients with endometrial cancer for future clinical trials with any targeted therapies. MMP1 expression was high in tissues and sera in patients with endometrial cancer. Lapatinib inhibited MMP1 via both HER2 and EGFR signaling pathways. Both AKT and ERK need to be inhibited for efficient MMP1 suppression by lapatinib. Activating protein-1 (AP-1) binding site of MMP1 promoter is uniquely required for MMP1 activation in endometrial cancer. Suppression of both c-fos and c-Jun bound to AP1 binding site is required for lapatinib inhibition.