Merck
  • Characterization and protective effect of a 29 kDa protein isolated from Coxiella burnetii by detergent Empigen BB.
Characterization and protective effect of a 29 kDa protein isolated from Coxiella burnetii by detergent Empigen BB.
European journal of epidemiology (1994-04-01)
M Lukácová, E Gajdosová, L Skultéty, E Kovácová, J Kazár
ABSTRACT
A 29 kDa protein, isolated from the outer membrane of Coxiella burnetii, strain Nine Mile phase I by detergent Empigen BB, was characterized. The failure in removing lipopolysaccharides (LPS) from preparations of the protein by the purification method used indicates a strong binding between proteins and LPS in the outer membrane of C. burnetii. The protein was immunogenic in mice and protected them against virulent C. burnetii challenge.
Sigma-Aldrich
EMPIGEN® BB detergent, ~30% active substance