Merck
  • NSD3, a new SET domain-containing gene, maps to 8p12 and is amplified in human breast cancer cell lines.
NSD3, a new SET domain-containing gene, maps to 8p12 and is amplified in human breast cancer cell lines.
Genomics (2001-05-26)
P O Angrand, F Apiou, A F Stewart, B Dutrillaux, R Losson, P Chambon
ABSTRACT
We describe the isolation and characterization of NSD3, the third member of a gene family including Nsd1 and NSD2. Murine Nsd1 was isolated in a search for proteins that interact with the ligand-binding domain of retinoic acid receptor alpha. NSD2 (also known as WHSC1 and MMSET) is located in the Wolf-Hirschhorn syndrome (WHS) critical region on 4p16.3 and is involved in multiple myeloma with t(4;14) translocations. The proteins Nsd1, NSD2, and NSD3 are highly similar within a block of about 700 amino acids. This block contains several conserved domains, such as the SET domain and the PHD finger, present in proteins involved in development and/or chromatin reorganization. The NSD3 gene consists of an 8.5-kb transcript composed of 23 coding exons and spans >90 kb of genomic DNA. NSD3 maps to chromosome band 8p12 and is amplified in several tumor cell lines and primary breast carcinomas.