Merck
  • Simple field assays to check quality of current artemisinin-based antimalarial combination formulations.
Simple field assays to check quality of current artemisinin-based antimalarial combination formulations.
PloS one (2009-10-01)
Jean-Robert Ioset, Harparkash Kaur
ABSTRACT
Malaria continues to be one of the major public health problems in Africa, Asia and Latin America. Artemisinin derivatives (ARTs; artesunate, artemether, and dihydroartemisinin) derived from the herb, Artemisia annua, are the most effective antimalarial drugs available providing rapid cures. The World Health Organisation (WHO) has recommended that all antimalarials must be combined with an artemisinin component (artemisinin-based combination therapy; ACT) for use as first line treatment against malaria. This class of drugs is now first-line policy in most malaria-endemic countries. Reports of ad hoc surveys from South East Asia show that up to 50% of the artesunate currently sold is counterfeit. Drug quality is rarely assessed in resource poor countries in part due to lack of dedicated laboratory facilities which are expensive to build, equip and maintain. With a view to address this unmet need we developed two novel colour reaction assays that can be used in the field to check the quality of ARTs. Our assays utilise thin layer chromatography silica gel sheets and 2, 4 dinitrophenylhydrazine or 4-Benzoylamino-2, 5-dimethoxybenzenediazonium chloride hemi (zinc chloride) salt as the reagents showing a pink or blue product respectively only in the presence ARTs. We are able to detect as low as 10% of ARTs in ACTs (WINTHROP--artesunate/amodiaquine, Coartem--artemether/lumefantrine and Duocortexcin--dihydroartemisinin/piperaquine). The assays have been validated extensively by testing eighty readily accessible and widely used drugs in malaria endemic countries. None of the other antimalarial drugs or a range of commonly used excipients, antiretroviral drugs or other frequently used drugs from the WHO essential drugs list such as analgesics or antibiotics are detected with our assays. Our two independent assays requiring no specialist training are specific, simple to use, rapid, robust, reproducible, inexpensive and, have successfully resulted in detecting two counterfeit drugs within a small scale screening survey of over 100 declared artemisinin-containing drugs collected from various Asian and African countries. These promising results indicate that the assays will provide a useful first test to assure the quality of the ACTs formulations in resource poor malaria endemic areas when there is an absence of dedicated medicines quality laboratory facilities.
Product Description
Supelco
2,4-Dinitrophenylhydrazine hydrochloric acid solution, for TLC derivatization, ~0.005 M in ethanol