Merck
  • Quantitative HPLC-MS analysis of nucleotide sugars in plant cells following off-line SPE sample preparation.
Quantitative HPLC-MS analysis of nucleotide sugars in plant cells following off-line SPE sample preparation.
Analytical and bioanalytical chemistry (2014-03-19)
Robert Behmüller, Ines C Forstenlehner, Raimund Tenhaken, Christian G Huber
ABSTRACT
An analytical workflow was developed for the absolute quantification of uridine diphosphate (UDP)-sugars in plant material in order to compare their metabolism both in wild-type Arabidopsis thaliana and mutated plants (ugd2,3) possessing genetic alterations within the UDP-glucose dehydrogenase genes involved in UDP-sugar metabolism. UDP-sugars were extracted from fresh plant material by chloroform-methanol-water extraction and further purified by solid-phase extraction with a porous graphitic carbon adsorbent with extraction efficiencies between 80 ± 5 % and 90 ± 5 %. Quantitative determination of the UDP-sugars was accomplished through HPLC separation with a porous graphitic carbon column (Hypercarb(TM)) which was interfaced to electrospray ionization Orbitrap mass spectrometry. The problem of instable retention times due to redox processes on the stationary phase were circumvented by grounding of the column effluent and incorporation of a column regeneration procedure using acetonitrile-water containing 0.10 % trifluoroacetic acid. The method was calibrated using external calibration and UDP as internal standard. Calibration functions were approximated by first- or second-order regression analysis for concentrations spanning three orders of magnitude. Upon injecting sample volumes of 2.65 μL, the limits of detection for the UDP-sugars were in the 70 nmol L(-1) range. Six different UDP-sugars, including UDP-glucose, UDP-galactose, UDP-arabinose, UDP-xylose, UDP-glucuronic acid, and UDP-galacturonic acid were found in concentrations of 0.4 to 38 μg/g plant material. Data evaluation by analysis of variance (ANOVA) revealed statistically significant differences in UDP-sugar concentrations between wild-type and mutant plants, which were found to conclusively mirror the impaired metabolic pathways in the mutant plants.
Product Description
Supelco
Supelclean ENVI-Carb SPE Tube, bed wt. 250 mg, volume 3 mL, pk of 54
Pricing and availability is not currently available.