Merck
  • EphA receptors regulate growth cone dynamics through the novel guanine nucleotide exchange factor ephexin.
EphA receptors regulate growth cone dynamics through the novel guanine nucleotide exchange factor ephexin.
Cell (2001-05-05)
S M Shamah, M Z Lin, J L Goldberg, S Estrach, M Sahin, L Hu, M Bazalakova, R L Neve, G Corfas, A Debant, M E Greenberg
ABSTRACT
Eph receptors transduce short-range repulsive signals for axon guidance by modulating actin dynamics within growth cones. We report the cloning and characterization of ephexin, a novel Eph receptor-interacting protein that is a member of the Dbl family of guanine nucleotide exchange factors (GEFs) for Rho GTPases. Ephrin-A stimulation of EphA receptors modulates the activity of ephexin leading to RhoA activation, Cdc42 and Rac1 inhibition, and cell morphology changes. In addition, expression of a mutant form of ephexin in primary neurons interferes with ephrin-A-induced growth cone collapse. The association of ephexin with Eph receptors constitutes a molecular link between Eph receptors and the actin cytoskeleton and provides a novel mechanism for achieving highly localized regulation of growth cone motility.