Merck
  • Determination of organophosphorus insecticides in natural waters using SPE-disks and SPME followed by GC/FTD and GC/MS.
Determination of organophosphorus insecticides in natural waters using SPE-disks and SPME followed by GC/FTD and GC/MS.
Fresenius' journal of analytical chemistry (2001-03-07)
D Lambropoulou, T Sakellarides, T Albanis
ABSTRACT
Two methods for the analysis of ten organophosphorus insecticides in natural waters using solid phase extraction disks containing C18 and SDB and solid phase microextraction fibers containing polyacrylate (PA) are developed. Bromophos ethyl, bromophos methyl, dichlofenthion, ethion, fenamiphos, fenitrothion, fenthion, malathion, parathion ethyl and parathion methyl were determined by GC/MS and GC/FTD. The SPE-disks require only 1000 mL of sample and provide a method limit of detection in the range of 0.01-0.07 microgram/L and recovery rates from 60.7 to 104.1%. The solid phase microextraction (SPME) technique requires 2-5 mL of water sample and provides a method limit of detection in the range of 0.01 to 0.05 microgram/L for all detectors and the recoveries compared to distilled water ranged from 86.2 to 119.7%. The proposed methods were applied to the trace level screening determination of insecticides in river water samples originating from different Greek regions.
Product Description
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Pricing and availability is not currently available.
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Pricing and availability is not currently available.