- Triclinic polymorph of dibenzotetra-thia-fulvalene.
Triclinic polymorph of dibenzotetra-thia-fulvalene.
Acta crystallographica. Section E, Structure reports online (2009-01-01)
Masashi Mamada, Yoshiro Yamashita
PMID21577501
ABSTRACT
Crystals of the title compound (DBTTF), C(14)H(8)S(4), feature a triclinic polymorph different from two known monoclinic polymorphs. In this form, there are two independent centrosymmetric half-mol-ecules in the asymmetric unit. Although the mol-ecular orientations are relatively similar to one of monoclinic polymorphs, the packing motif is different.