Merck
  • Diagnostic utility of FLI-1 monoclonal antibody and dual-colour, break-apart probe fluorescence in situ (FISH) analysis in Ewing's sarcoma/primitive neuroectodermal tumour (EWS/PNET). A comparative study with CD99 and FLI-1 polyclonal antibodies.
Diagnostic utility of FLI-1 monoclonal antibody and dual-colour, break-apart probe fluorescence in situ (FISH) analysis in Ewing's sarcoma/primitive neuroectodermal tumour (EWS/PNET). A comparative study with CD99 and FLI-1 polyclonal antibodies.
Histopathology (2006-12-14)
P Mhawech-Fauceglia, F Herrmann, R Penetrante, A Beck, S Sait, A M Block, K Odunsi, J Fisher, L Balos, R T Cheney
ABSTRACT
To compare the sensitivity and specificity of the recently commercially available FLI-1 monoclonal (FLI-1m) antibody with the currently used antibodies [CD99 and FLI-1 polyclonal (FLI-1p)] in the diagnosis of Ewing's sarcoma/primitive neuroectodermal tumour (EWS/PNET) and to determine the diagnostic value of the EWSR1 (22q12) dual-colour, break-apart rearrangement probe fluorescence in situ hybridization (FISH) technique. Forty-three cases of well-documented EWS/PNET and 15 non-EWS/PNET cases were retrieved from the archival files. Immunohistochemistry (IHC) for FLI-1p, FLI-1m and FISH analysis was performed. The most sensitive and specific test panel for the diagnosis of EWS/PNET is the combination of CD99 and FLI-1p. FISH had a very high specificity (100%) but only a moderate sensitivity (50%). The combination of CD99 and FLI-1p is the method of choice for the diagnosis of EWS/PNET. EWRS1 (22q12) dual-colour, break-apart rearrangement probe FISH should be used as a confirmatory test in addition to CD99 and FLI1-p due to its high specificity.